missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva HyClone™ Dulbecco's Modified Eagles Medium

Designed to support cell culture growth and biomanufacturing processes.

Supplier:  Cytiva SH3008101

 View more versions of this product

Catalog No. SH3008101



Description

Description

  • Format: Liquid
Specifications

Specifications

ISO 9001:2000
Liquid
0.1 μm Sterile-filtered
No Glutamine, No Sodium Pyruvate
2° to 8°C, away from light
DMEM/High glucose without L-glutamine, and sodium pyruvate
Glucose
500 mL
SDS
Documents

Documents

Promotions

Promotions