Gibco™ Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (0.5 x 106 cells)
Human Umbilical Vein Endothelial Cells, HUVEC Pooled, (0.5 x 106 cells)
Supplier: Gibco™ C0155C
Description
Primary human umbilical vein endothelial cells, pooled from multiple donors, cryopreserved at the end of the primary culture.Specifications
Liquid nitrogen temperature | |
Endothelial Cells (Umbilical Vein) | |
Cryopreserved | |
Pooled | |
Cascade Biologics™ | |
Hepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1 |
Newborn (14 days or less) | |
Adherent Cell Culture | |
Human | |
≥500000 | |
Human Umbilical Vein Endothelial Cells |
For Research Use Only. Not for use in diagnostic procedures.