Gibco™ Human Epidermal Keratinocytes, adult, Animal Product-Free (HEKa-APF) DFS Item
Human Epidermal Keratinocytes, adult, Animal Product-Free (HEKa-APF)
Manufacturer:  Gibco™ LSC0215C
Catalog No. C0215C
Description
Description
Primary human epidermal keratinoctyes from adult skin. Isolated, grown and cryopreserved at the end of the primary culture in an animal product-free environment.
  • Viability at least 70%
  • ≥500,000 viable cells⁄vial
  • Capable of at least 25 population doublings
  • Mycoplasma: Not Detected
  • Hepatitis B: Not Detected
  • Hepatitis C: Not Detected
  • HIV-1: Not Detected
  • Bacteria, yeast, and other fungi: Not Detected
  • Specifications
    Specifications
    Liquid Nitrogen
    1 Vial(s)
    Single donor
    Liquid Nitrogen
    Primary cell culture
    Gibco Human Epidermal Keratinocytes, 1 Vial, Cryopreserved, Human Species, Adult Age, Keratinocytes Cell Type, Single Donor Source, Liquid Nitrogen Storage, For Primary Cell Culture
    Keratinocytes
    Human Epidermal Keratinocytes
    Adherent cell culture
    Documents
    For Research Use Only. Not for use in diagnostic procedures.