Troemner™ Hooked Stainless-Steel Weight Sets and Individual Weights
Weights can be hooked together at top and bottom.
Supplier: Troemner™ 30390790
Description
The design of the hook weight allows the bottom hook to be recessed in the bottom so the hook weights can be set on a flat surface.
- Adjusted to tolerances specified by ANSI™/ASTM™ Class 7
- Weight sets are supplied in an open plastic storage case
Specifications
Metric Hook Weight | |
Stainless Steel | |
7.85g/cm3 at 20°C |
ANSI/ASTM Class 7 | |
50 g |