Hoefer™ Glass Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
Supplier:  Hoefer™ PFRSE6102
 View more versions of this product
Catalog No. 36-100-3845
Description
Description
  • For use with SE660, SE600X Chroma, SE600, SE640, SE410, and SE400 Vertical Electrophoresis Units
  • Regular (straight, without notch)
  • Supplied in packs of 2
Specifications
Specifications
SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
Glass
18 x 16 cm
16 cm
Glass Plate
2/Pk.
18 cm