missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

High Vacuum Grease

Seal and lubricate vacuum and pressure systems.

Manufacturer:  Fisher Scientific 1597418EMD

Catalog No. S41718




Description

Description

  • Low volatility to hold deep vacuums
  • Resists oxidation, melting and gumming
  • Good chemical and thermal stability
  • Maintains consistency at temperatures from -40° to +204°C (-40° to +400°F)
  • Capacity: 5.3 oz. (150.2g) tube

Specifications

Specifications

High Vacuum Grease
Each
-40° to 204°C
Used for sealing vacuum and pressure systems
-40° to 400°F
Documents