missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ High Performance Bench Chemical Fume Hood

Fisherbrand™ High Performance Chemical Fume Hoods are innovative and purpose driven products for the research and healthcare industries.

Specifications

Certifications/Compliance UL 1805
Color Fisherbrand White
Depth (English) 35.875 in.
Electrical Requirements 120 Volt, 20 Amp
Flow Rate 80 to 120 FPM
View More Specs

Products 4
Catalog Number Mfr. No. Width (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Price Quantity  
06000584 View Documents Fisherbrand™
F35048G7
48 in. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06000585 View Documents Fisherbrand™
F35060G7
60 in. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06000586 View Documents Fisherbrand™
F35072G7
72 in. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
06000587 View Documents Fisherbrand™
F35096G7
96 in. N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • The expansive interior is enhanced by a 43 in. tall sight line with a 17 in. high glass observation panel.
  • Includes frameless vertical rising sash with a 24 in. clear sash opening.
  • Easy conversion from CAV to VAV use
  • Chain and Sproket sash mechanism for ease in operation
  • Tempered glass sash with full-length formed steel handle for a neat, clean appearance and streamline air passage.
  • Stainless steel round duct collar(s)
  • Color: Fisherbrand White
  • Fisherbrand hoods are UL 1805 listed and are tested in accordance to ASHRAE-110.
  • All hoods are pre-wired to a junction box on the roof of the hood with two 120 volt AC 20 amp GFCI receptacles on the side posts, T8-fluorescent light fixture with bulb, 20 amp light switch and a duplex for an alarm on the roof
  • Each front post and interior end liner is punched and capped for up to five (5) remote control service fittings
  • The left hand post is punched and covered for a second electrical fixture at the top which may be used for a fan switch or other electrical device. The right post is punched and covered for an alarm.
  • Service fittings, fan switch, work top, cup sink, alarm, and base units must be ordered separately.
Specifications

Specifications

UL 1805
35.875 in.
80 to 120 FPM
57.125 in.
T8-Fluorescent Light Fixture and Bulb
24 in.
Fisherbrand White
120 Volt, 20 Amp
48 in.
60 Hz
Polyglass Liner
120 Volt