HF Scientific™ DRT-15CE Portable Turbidimeters Reusable Glass Cuvettes and Stand
For grab samples
$277.56 - $939.79
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
15-393-4
|
HF Scientific™
HF50052 |
Cuvette | 10 |
Pack of 10 Each for $939.79
|
|
||||
15-393-5
|
HF Scientific™
HF50051 |
Cuvette | 3 |
Pack of 3 Each for $343.15
|
|
||||
15-393-6
|
HF Scientific™
HF19981 |
Cuvette Stand | 1 |
Each for $277.56
|
|
||||
Description
- Reusable 28 × 70mm glass cuvets
- Hard-tempered glass surface resists surface scratching
- Supplied with threaded light shield caps
- Cuvet stand stores up to 11 cuvets or calibration standards
Specifications
DRT 15C Portable Turbidimeter |