missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexanes, for residue analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 610180040
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For residue analysisEcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Specifications
Hexanes | |
0.6590g/mL | |
Conforms to reference | |
C6H14 | |
MFCD00009520 | |
14, 4694 | |
86.18 |
92112-69-1,110-54-3 | |
4 L | |
EcoSafPak™ | |
CH3(CH2)4CH3 | |
01, 142 | |
Hex | |
Residue Analysis |
RUO – Research Use Only