missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexanes, Optima™, Fisher Chemical

High purity and versatile solvent suitable for HPLC, spectrophotometry, GC/MS, and pesticide residue analysis

$242.15 - $407.08

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
H303-1 SDS View Documents Thermo Scientific
H3031
1L Glass Bottle Each for $242.15
Case of 6 Each for $1,452.90
H303SK-4 SDS View Documents Thermo Scientific
H303SK4
4L Glass Bottle Each for $407.08
Case of 4 Each for $1,628.32
H303-4 SDS Accessories View Documents Thermo Scientific
BPH3034
4L Glass Bottle Each for $312.23
Case of 4 Each for $1,248.92
Description

Description

  • Mixture of hexanes where n-hexane content is ≥60%
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis and tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Chemical Identifiers

110-54-3
86.178
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

1 AU max. at 195nm, 0.25 AU max. at 210nm, 0.07 AU max. at 220nm, 0.005 AU max. at 254nm
Vapor Pressure: 160mbar at 20°C
C6H14
1208
99.9 % min.
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
High purity mobile phase for HPLC and/or extraction solvent for GC applications
1ppb max.
Glass Bottle
1.3740 to 1.3780
Pass test (GC-MS)
Pass test
0.01% Max.
−95°C (−139°F)
Certificate of Lot Analysis included with each bottle; UV and other chromatograms available on request.
Colorless
1L
110-54-3
99.9% min by GC
MFCD02179311
60 % min.
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.178
CHEBI:29021
Liquid
0.659g/cm³
Pass test
10ng/L max.
1ppm max.
0.005 % max.
160mbar at 20°C
0.0003meq/g max.
0.31 mPa/s at 20°C
69°C
-95°C
Hexanes
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Extremely flammable liquid and vapor. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of impaired fertility. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid