missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexanes, 99%, ACS reagent, mixture of isomers, Thermo Scientific Chemicals
$111.18 - $630.06
Specifications
Chemical Name or Material | Hexanes |
---|---|
Melting Point | -95.0°C |
Name Note | Mixture of isomers |
CAS | 92112-69-1 |
Density | 0.6590g/mL |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC423765000
|
Thermo Scientific Chemicals
423765000 |
500 mL | EcoSafPak™ Glass Bottle |
Each for $111.18
Case of 6 Each for $667.08
|
|
||||
AC423760010
|
Thermo Scientific Chemicals
423760010 |
1 L | EcoSafPak™ Glass Bottle |
Each for $141.80
|
|
||||
AC423760025
|
Thermo Scientific Chemicals
423760025 |
2.5 L | Glass Bottle |
Each for $187.40
|
|
||||
AC423760100
|
Thermo Scientific Chemicals
423760100 |
10 L | Plastic Drum |
Each for $630.06
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexanes | |
Mixture of isomers | |
0.6590g/mL | |
−22°C | |
1.3748 to 1.3810 | |
0.001% max. | |
C6H14 | |
CH3(CH2)4CH3 | |
Vapor Pressure: 160mbar at 20°C | |
01, 142 | |
Hex | |
86.18 | |
ACS Reagent |
-95.0°C | |
92112-69-1 | |
69.0°C | |
Liquid | |
99% | |
0.659 | |
0.0003 meq/g max. | |
MFCD00009520 | |
0.31 mPa.s (20°C) | |
14, 4694 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform | |
99% |
RUO – Research Use Only