missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexagram™ Weights

Explore weight and measurement with these easily stackable Hexagram Weights.

Manufacturer:  Fisher Science Education LER750

Catalog No. S40446


 

Alert:

CHOKING HAZARD (1). Small parts. Not for children under 3 yrs.


Includes: Set of 54: twenty 1g, twenty 5g, ten 10g and four 20g weights.

Specifications

Description

Set, Weight; Hexagram; Set of 54: twenty 1g, twenty 5g, ten 10g and four 20g weights; Grades 1-5.

  • Offer visual cues in measurement activities

Order Info

Grades 1 to 5

Description & Specifications

Specifications

Hexagram weight set
Weight: 20 x 1g, 20 x 5g, 10 x 10g and 4 x 20g