missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Heracell™ VIOS 160i Tri-Gas CO2 Incubators

Achieve your next breakthrough. This CO2 incubator delivers the performance reliability, ease of operation, and value required to support a range of culturing needs.

Specifications

Capacity (English) Shelf 22/66 lb.
Capacity (Metric) 255L
Capacity (Metric) Shelf 10/30kg
Certifications/Compliance CE, CSA
CO2 Concentration Control ±0.1°C
View More Specs

Products 4
Catalog Number Mfr. No. Description CO2 Sensor Technology Capacity (English) Cabinet Material Price Quantity  
Catalog Number Mfr. No. Description CO2 Sensor Technology Capacity (English) Cabinet Material Price Quantity  
13998236 View Documents Thermo Scientific™
51031048
Heracell VIOS 250i Tri-Gas C02 incubator single chamber TC sensor 9.0 cu. ft. 18 gauge, cold-rolled steel, powder coated N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-998-237 View Documents Thermo Scientific™
51031047
Heracell VIOS 250i Tri-Gas C02 incubator single chamber TC sensor 9.0 cu. ft. 18 gauge, cold-rolled steel, powder coated N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13998238 View Documents Thermo Scientific™
51031159
Heracell VIOS 250i Tri-Gas C02 incubator single chamber IR sensor 9.0 cu. ft. 18 gauge, cold-rolled steel, powder coated N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13998239 View Documents Thermo Scientific™
51031160
Heracell VIOS 250i Tri-Gas C02 incubator single chamber IR sensor 9.0 cu. ft. 18 gauge, cold-rolled steel, powder coated N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Achieve your next breakthrough. The Thermo Scientific™ Heracell™ VIOS CO2  incubator is specifically designed to deliver the performance reliability, ease of operation, and value required to support a range of culturing needs from basic research to the demanding, leading-edge applications, so you're ready for whatever comes next.

Better solutions for optimal cell growth. Get added culturing flexibility with our CO2 incubators with variable oxygen control (tri-gas).

Direct Heat Chamber
  • 165L (5.8 cu ft) electropolished stainless steel chamber with compact footprint, easily stackable
  • Standard left hand door swing, with inner 3 segmented gas tight glass doory
THRIVE Active Airflow
  • In-chamber fan gently distributes clean, humidified air throughout the chamber ensuring homogeneous conditions
  • Fast recovery of all parameters ten minutes or less for stable culturing conditions
  • Auto-stop function disables fan operation during door openings to minimize air exchange
ISO 5 HEPA Filtration
  • In-chamber HEPA continuously filters the entire chamber air volume every 60 seconds
  • ISO 5 clean room air quality is achieved in the culture chamber within five minutes
  • Continuous protection by minimizing ability for contaminants to settle on interior surfaces
Protected Humidity Reservoir
  • Directly heated water reservoir provides stable high relative humidity levels, preventing culture desiccation
  • Cover limits particles and spilled media from settling into the reservoir
  • Condensation free inner chamber, prevents a breeding ground for contaminant
  • 5X faster humidity recovery than traditional water pan designs
  • For easier water handling, humidity reservoir may be filled or drained without the removal of shelves or cultures
In situ sensors and probes
  • Dual temperature probes for over temperature protection with operational back up
  • Choice of thermal conductivity (TC) with rh compensation or single beam, dual wavelength infrared (IR) CO2 sensor
  • O2 control ranges 1 to 21% (hypoxic) and 5 to 90% (hyperoxic)
Steri-Run, high temperature disinfection cycle
  • Overnight cycle simplifies cleaning and eliminates the need for separate autoclaving of parts
  • 180°C cycle assures uniform 6-log sterilization on all chamber surfaces eliminating biological contaminants
Intuitive, interactive iCAN™ Touchscreen Control
  • Convenient on-screen user prompts and reminders for easy navigation
  • Selectable languages simplify operation: English, Spanish, German, French, Italian, Japanese and Mandarin
  • Visibility to changes in culture environment on performance graphs and error and data logs
Ordering Information:

For 220V Tri-gas models, please order a Heracell VIOS single base model (51030287, 51030286, 51030478 or 51030476) with the factory installed O2 controlled option
Specifications

Specifications

22/66 lb.
10/30kg
±0.1°C
1 to 20%
24.8 in.
Under 12 hours
26.4 in.
670mm
19.68 in.
215 lb.
>/=93%
3°C above ambient to 55°C
30.5 in.
22.1 in.
607mm
120V 50/60Hz
36.8 in.
3 standard/12 maximum
75W during incubation, 125W during sterlization
255L
CE, CSA
6.3A during incubation, 10.4A during sterilization
USB, and optional 4-20mA
629mm
38.1 in.
968mm
Integrated reservoir
500mm
97.5kg
3 standard/12 maximum
120V
23.9 in.
774mm
560mm
50/60Hz
934mm
42mm access port, rear left