missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 172801000

 View more versions of this product

Catalog No. AC172801000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Heptafluorobutyric acid
-17.5°C
1.6400g/mL
Authentic
Glass bottle
1.3
MFCD00004171
1.64
Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
214.04
CHEBI:39426
99%
375-22-4
Colorless to Yellow
120.0°C (755.0 mmHg)
99%
C4HF7O2
CF3CF2CF2CO2H
100 g
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
YPJUNDFVDDCYIH-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutanoic acid
9777
214.04
Liquid

Chemical Identifiers

375-22-4
214.04
YPJUNDFVDDCYIH-UHFFFAOYSA-N
9777
2,2,3,3,4,4,4-heptafluorobutanoic acid
C4HF7O2
MFCD00004171
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech
CHEBI:39426
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 206-786-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only