missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free
$303.32 - $303.32
Chemical Identifiers
CAS | 7365-45-9 |
---|---|
Molecular Formula | C8H18N2O4S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00006158 |
InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
Synonym | 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid |
PubChem CID | 23831 |
ChEBI | CHEBI:42334 |
IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC329850500
|
Thermo Scientific Chemicals
329850500 |
50 g |
Each for $303.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7365-45-9 | |
238.30 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
23831 | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
C8H18N2O4S | |
MFCD00006158 | |
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid | |
CHEBI:42334 | |
OCCN1CCN(CCS(O)(=O)=O)CC1 |
Specifications
(0.5 M in water), (0.5 M in water) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm | |
234.0°C to 238.0°C | |
White | |
Crystalline Powder | |
99+% | |
0.5% max. (105°C) | |
MFCD00006158 | |
23, V,2, 379 | |
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
23831 | |
238.3 | |
Molecular Biology |
(0.5M in water) | |
7365-45-9 | |
5 to 6.5 (0.5 M aq. soln.) | |
50 g | |
Authentic | |
C8H18N2O4S | |
Plastic bottle | |
15, 4689 | |
Solubility in water: 704g/L (20°C). | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
≥99% | |
HEPES free acid |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 230-907-9
RTECSNumber : TL6809000
TSCA : TSCA
RUO – Research Use Only