missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HEPES, free acid, >98%, MP Biomedicals™
$72.15 - $3236.23
Chemical Identifiers
CAS | 7365-45-9 |
---|---|
Molecular Formula | C8H18N2O4S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00006158 |
InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
Synonym | hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate |
PubChem CID | 23831 |
ChEBI | CHEBI:42334 |
SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10192625
|
MP Biomedicals Inc
0210192625 |
25 g |
Each for $72.15
|
|
|||||
ICN10192650
|
MP Biomedicals Inc
0210192650 |
50 g |
Each for $114.13
|
|
|||||
ICN10192680
|
MP Biomedicals Inc
0210192680 |
100 g |
Each for $134.38
|
|
|||||
ICN10192683
|
MP Biomedicals Inc
0210192683 |
250 g |
Each for $315.46
|
|
|||||
ICN10192690
|
MP Biomedicals Inc
0210192690 |
500 g |
Each for $527.24
|
|
|||||
ICN10192691
|
MP Biomedicals Inc
0210192691 |
1 kg |
Each for $960.28
|
|
|||||
ICN10192694
|
MP Biomedicals Inc
0210192694 |
5 kg |
Each for $3,236.23
|
|
|||||
Chemical Identifiers
7365-45-9 | |
238.30 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
23831 | |
OCCN1CCN(CCS(O)(=O)=O)CC1 |
C8H18N2O4S | |
MFCD00006158 | |
hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate | |
CHEBI:42334 |
Specifications
7365-45-9 | |
>98% | |
MFCD00006158 | |
hepes, 2-4-2-hydroxyethyl piperazin-1-yl ethanesulfonic acid, 4-2-hydroxyethyl-1-piperazineethanesulfonic acid, 1-piperazineethanesulfonic acid, 4-2-hydroxyethyl, monosodium salt, hepes, 4-2-hydroxyethyl piperazine-1-ethanesulfonic acid, unii-rww266ye9i, 4-2-hydroxyethyl-1-piperazineethane sulfonic acid, n-2-hydroxyethylpiperazine-n'-ethanesulfonate | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
Powder |
White | |
C8H18N2O4S | |
25 g | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
23831 | |
238.3 | |
HEPES |
Safety and Handling
EINECSNumber : 230-907-9
Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.