missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Hematocrit Rotor

Process capillary tubes via centrifugation with the Thermo Scientific™ Hematocrit Rotor.

Manufacturer:  Thermo Scientific™ 75003473

Catalog No. 05-375-105



Description

Description

Process capillary tubes via centrifugation with the Thermo Scientific™ Hematocrit Rotor.

Features:
  • Holds 24 hematocrit tubes
  • Max. Speed/RCF: 14,800rpm/16,800 x g
Includes: Manual count reader card
Specifications

Specifications

Hematocrit Rotor
24 Capillaries
14,800rpm
2.0cm
Sorvall Legend Micro 17/17R/21/21R, Heraeus Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
Aluminum
16,800 x g
8.5cm
No
90°
Documents

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.