missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hamilton™ 800 Microliter™ Syringes, N Termination

Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 µL up to 250 µL.

$142.06 - $217.02

Specifications

Connection Type Cemented
Max. Pressure 2000psi
Needle Point Style Sharp, Bevel, Curved, Non-coring
Min. Temperature (English) 50°F
Autoclavable No
View More Specs

Products 5 Description

Description

  • Ideal for dispensing volumes from 0.5 μL up to 250 μL
  • A variety of different terminations and needle options are available
  • Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending
Specifications

Specifications

Cemented
Sharp, Bevel, Curved, Non-coring
No
51mm
Stainless Steel
2000psi
50°F
2 in.
Borosilicate Glass
Microliter™ Syringe