Hamilton™ 700 7000/1700 1000 Series Microliter™/Gastight™: TLL Termination
Syringes have built-in Chaney Adapters
$246.74 - $404.77
Specifications
Needle Point Number No Needle Included
View More Specs
Products 4
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Price Quantity  
14-815-126
View Documents Promotion Details
Hamilton™
HAM81324
1 mL
Each for $275.75
14-815-151
View Documents Promotion Details
Hamilton™
HAM81424
2.5 mL
Each for $399.94
14-815-166
View Documents Promotion Details
Hamilton™
HAM81524
5 mL
Each for $246.74
14-815-182
View Documents Promotion Details
Hamilton™
HAM81624
10 mL
Each for $404.77
Description
Description
Built to adjust reproducible injections and to prevent the plunger from bending. Syringes in this category are either Microliter (700/7000 series) or Gastight (1700/1000 series) syringes. Microliter syringes have a stainless steel plunger that is hand-fitted to the glass barrel and are designed for liquid sampling. Gastight syringes have a PTFE plunger tip that creates tight connection between the syringe barrel and plunger allowing for liquid or gas sampling. These syringes have PTFE luer lock termination. Available in 0.5μL to 10mL in volume.
PTFE Luer Lock (TLL) – This is a male luer with a nickel plated brass hub. This termination accepts Metal Hub and Kel-F Hub needles and locks them in place
Needle Point Styles
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 26s, 25, 24, 23, 22, 22s
  • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 26s, 25, 24, 23, 22, 22s
Specifications
Specifications
No Needle Included
SDS
missing translation for 'documents'
missing translation for 'documents'