GVS Nitrocellulose-Mixed Esters of Cellulose Membrane Filters
Uniform microporous structure has low extractables, low ash
$218.75 - $810.88
Specifications
Material | MCE |
---|---|
For Use With (Application) | Microbiological determinations; industrial hygiene procedures; clarifying aqueous solutions; determination of lead, chromium, cadmium, beryllium, arsenic, airborne asbestos and other particulates. |
Hydrophobicity | Hydrophilic |
Catalog Number | Mfr. No. | Color | Diameter (Metric) | Pore Size | Membrane Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Diameter (Metric) | Pore Size | Membrane Type | Price | Quantity | ||
E02WP02500
|
GVS
1214898 |
White | 25 mm | 0.2 μm | Plain |
Pack of 100 Each for $261.69
|
|
||
E02WP09025
|
GVS
1214941 |
White | 90 mm | 0.2 μm | Plain |
Pack of 25 Each for $218.75
|
|
||
E02WP01300
|
GVS
1214882 |
White | 13 mm | 0.2 μm | Plain |
Pack of 100 Each for $232.41
|
|
||
E02WP04700
|
GVS
1214909 |
White | 47 mm | 0.2 μm | Plain |
Pack of 100 Each for $221.34
|
|
||
E02WP14225
|
GVS
1214950 |
White | 142 mm | 0.2 μm | Plain |
Pack of 25 Each for $401.81
|
|
||
E04BG04700
|
GVS
1214977 |
Black | 47 mm | 0.5 μm | Gridded |
Pack of 100 Each for $492.63
|
|
||
E04WG04700
|
GVS
1215207 |
White | 47 mm | 0.5 μm | Gridded |
Pack of 100 Each for $374.75
|
|
||
E04WP02500
|
GVS
1215263 |
White | 25 mm | 0.5 μm | Plain |
Pack of 100 Each for $276.84
|
|
||
E04WP09025
|
GVS
1215305 |
White | 90 mm | 0.5 μm | Plain |
Pack of 25 Each for $247.53
|
|
||
E04WP14225
|
GVS
1215316 |
White | 142 mm | 0.5 μm | Plain |
Pack of 25 Each for $467.66
|
|
||
E06WP04700
|
GVS
1215380 |
White | 47 mm | 0.7 μm | Plain |
Pack of 100 Each for $397.34
|
|
||
E08BG04700
|
GVS
1215412 |
Black | 47 mm | 0.8 μm | Gridded |
Pack of 100 Each for $428.78
|
|
||
E08WP03700
|
GVS
1215426 |
White | 37 mm | 0.8 μm | Plain |
Pack of 100 Each for $341.78
|
|
||
E08BP02500
|
GVS
1215415 |
Black | 25 mm | 0.8 μm | Plain |
Pack of 100 Each for $394.13
|
|
||
E08WG04700
|
GVS
1215421 |
White | 47 mm | 0.8 μm | Gridded |
Pack of 100 Each for $484.59
|
|
||
E08WP02500
|
GVS
MM1215425 |
White | 25 mm | 0.8 μm | Plain |
Pack of 100 Each for $276.84
|
|
||
E08WP04700
|
GVS
1215428 |
White | 47 mm | 0.8 μm | Plain |
Pack of 100 Each for $397.34
|
|
||
E08BP04700
|
GVS
1215416 |
Black | 47 mm | 0.8 μm | Plain |
Pack of 100 Each for $428.78
|
|
||
E08WP09025
|
GVS
1215431 |
White | 90 mm | 0.8 μm | Plain |
Pack of 25 Each for $387.66
|
|
||
E12WG04700
|
GVS
1215437 |
White | 47 mm | 1.2 μm | Gridded |
Pack of 100 Each for $378.84
|
|
||
E12WP02500
|
GVS
1215440 |
White | 25 mm | 1.2 μm | Plain |
Pack of 100 Each for $251.69
|
|
||
E12WP09025
|
GVS
1215442 |
White | 90 mm | 1.2 μm | Plain |
Pack of 25 Each for $302.50
|
|
||
E12WP04700
|
GVS
1215441 |
White | 47 mm | 1.2 μm | Plain |
Pack of 100 Each for $453.72
|
|
||
E50WP02500
|
GVS
1215450 |
White | 25 mm | 5 μm | Plain |
Pack of 100 Each for $251.69
|
|
||
E50WP04700
|
GVS
1215451 |
White | 47 mm | 5.0 μm | Plain |
Pack of 100 Each for $434.06
|
|
||
E04WP04700
|
GVS
1215281 |
White | 47 mm | 0.5 μm | Plain |
Pack of 100 Each for $351.75
|
|
||
E01WP04700
|
GVS
1214533 |
White | 47 mm | 0.1 μm | Plain |
Pack of 100 Each for $810.88
|
|
||
Description
- Cellulose nitrate/cellulose acetate polymers offer fast flow, high throughput, no contaminating fibers
- No cytotoxic wetting agents
- Hydrophilic; extractables <4%; biologically inert; <0.05% ash
- Gridded types for particle counting
- Thickness, 150μm (except 0.10μm membranes, 130μm)
- 0.22 and 0.45μm sizes are tested for microbial recovery, bacteria retention
- White membranes autoclavable at 121°C (250°F) for 20 minutes
- Black membranes provide dark background for particle analyses
- Not autoclavable
Microbiological determinations; industrial hygiene procedures; clarifying aqueous solutions; determination of lead, chromium, cadmium, beryllium, arsenic, airborne asbestos and other particulates.
Order Info
In dispenser packs of 25 or 100. Nitrocellulose 47mm membrane filters in 0.22 and 0.45μm thickness are also available in presterilized packs, as well as autoclavable packs suitable for onsite presterilization. Order online or contact your Fisher Scientific™ Customer Service Representative.
Specifications
MCE | |
Microbiological determinations; industrial hygiene procedures; clarifying aqueous solutions; determination of lead, chromium, cadmium, beryllium, arsenic, airborne asbestos and other particulates. | |
Hydrophilic |