missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Graduated Beaker Set
Unbreakable plastic.
Supplier: Customer Channels Group (Ccg) LER306
Alert:
CHOKING HAZARD—Small parts. Not for children under 3 yrs.
Description
- Set of five beakers
- Wide mouths make measuring and pouring easy
- Pieces nest for convenient storage
- Set includes one each of 50mL, 100mL, 250mL, 500mL and 1L containers
- Grades: Pre-K+
Specifications
50 mL, 100 mL, 250 mL, 500 mL, and 1000 mL | |
50, 100, 250, 500, 1000 mL | |
Yes |
Plastic | |
Beaker |