missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ Assembled HDPE Round Bottles, Red Cap with Wad
Easier to handle, require less space (13-20%) on the shelf and are ideal for sampling and storage. Corning™ HDPE Round Bottles are lightweight and break resistant
Supplier: Corning™ LR150A14
Description
- Break-resistant and translucent high-density polyethylene (HDPE)
- HDPE cap
Compliance
Food contact compliant
Specifications
Round Bottle | |
Wad | |
Red | |
Clinical, Industry, Microbiology | |
108 mm | |
Sterile | |
Raw material is compliant with regulation EC N°1935/2004 Sterility Assurance Level based on ISO 11137 : STERILE SAL 10-3 |
Screw Cap | |
150 mL | |
Sampling and Storage Bottle | |
HDPE | |
250/ Cs. | |
Red |