missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Glucose-Glutamic Acid Solution, Standard Check Solution for Biochemical Oxygen Demand (BOD), Ricca Chemical

$59.94 - $393.64

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
325516 View Documents Ricca Chemical Company
RI325516
500mL Amber Glass Each for $114.03
3255246 SDS View Documents Ricca Chemical Company
RI3255246
144mL Glass vial Pack of 24 Each for $203.94
325532 View Documents Ricca Chemical Company
RI325532
1L Amber Glass Each for $158.39
32554 SDS View Documents Ricca Chemical Company
RI32554
120mL Amber Glass Each for $59.94
3255486 SDS View Documents Ricca Chemical Company
RI3255486
288mL Glass vial Pack of 48 Each for $393.64
Specifications

Chemical Identifiers

50-99-7
180.156
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
GZCGUPFRVQAUEE-SLPGGIOYSA-N
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O

Specifications

50-99-7,56-86-0,7732-18-5
100.0,0.02,0.02
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
Liquid
Colorless
Glucose-Glutamic Acid Solution, Standard Check Solution for Biochemical Oxygen Demand (BOD)
98.0,0.02,0.01
C6H12O6
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
Amber Glass
500mL