Thermo Scientific™ GLD Pro Gas Leak Detector
Quickly locate and identify gas leaks in your GC instrument with the Thermo Scientific™ GLD Pro Gas Leak Detector.
Manufacturer:  Thermo Scientific™ 66002001
Catalog No. 66002-001
Description
Description
Detect and identify gas leaks quickly with the Thermo Scientific™ GLD Pro Gas Leak Detector. Specifically designed for use with GC systems, it detects minute leaks of a wide range of laboratory gases. This hand-held, easy-to-use detector features both visual and audible alarms and utlilizes an LED light display to indicate the intensity of the leak. The small probe detector is designed to reach difficult and confined locations.
Additional Product Information
  • Pushbutton on/off switch
  • Pushbutton zero function
  • Automatic shutoff (five minutes)
  • Rechargable battery (up to six hours operation)
  • Durable storage case
  • Probe holder
  • One-year warranty
Specifications
Specifications
GC
Gas Leak Detector
GLD Pro Gas Leak Detector; Auto shutoff; 6-hr rechargable battery
Documents