Gilson™ TRACKMAN™ Duo-plate Pipeting Tracker
TRACKMAN™ Duo-plate Pipeting Tracker with a small footprint will increase productivity and eliminate potential errors.
Supplier: Gilson™ F70301
Description
- Easily switch from using one size plate to another on the same instrument
- Timer function enables hands-free operation and optional foot pedal is available for hands-free control
- Long lasting battery allows cordless operation for up to eight hours so you can take your TRACKMAN anywhere
- Duo-plate tracker lights the way for pipetting to avoid losing track when pipetting expensive samples
- Can be used with both 96 and 384 well microplates, and single and multichannel pipettes
Specifications
All Pipettes with Tips Mounted | |
16 | |
Long lasting battery |
Polypropylene | |
96 and 384 Well microplates, single and multichannel Pipette |