Gilson™ EXPERT™ Universal Fit Standard Pipette Tips
Feature patented, low-retention polymer that creates a hydrophobic surface to minimize sample binding while dispensing
$5.28 - $86.14
Specifications
Description | Standard Pipette Tips |
---|---|
Color | Clear |
Product Line | EXPERT |
Material | Polypropylene |
Catalog Number | Mfr. No. | Graduations | Length (Metric) | Volume (Metric) | Model | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Graduations | Length (Metric) | Volume (Metric) | Model | Price | Quantity | ||
F1731001G
|
Gilson™
F1731001 |
2 and 10μL | 2.883cm | 0.1 to 10 μL | E10 Tipack |
Pack of 960 Each for $73.84
|
|
||
F1731011G
|
Gilson™
F1731011 |
2 and 10μL | 2.883cm | 0.1 to 10 μL | E10ST Sterilized Tipack |
Pack of 960 Each for $83.06
|
|
||
F1732001G
|
Gilson™
F1732001 |
2, 5, 10μL | 4.534cm | 0.1 to 10 μL | EL10 Tipack |
Pack of 768 Each for $73.84
|
|
||
F1732011G
|
Gilson™
F1732011 |
2, 5, 10μL | 4.534cm | 0.1 to 10 μL | EL10ST Sterilized Tipack |
Pack of 768 Each for $83.06
|
|
||
F1733001G
|
Gilson™
F1733001 |
10, 50, 100μL | 5.144cm | 1 to 200 μL | E200 Tipack |
Pack of 768 Each for $73.84
|
|
||
F1733011G
|
Gilson™
F1733011 |
10, 50, 100μL | 5.144cm | 1 to 200 μL | E200ST Sterilized Tipack |
Pack of 768 Each for $86.14
|
|
||
F1734511G
|
Gilson™
F1734511 |
2, 5, 10μL | 4.534cm | 0.1 to 10 μL | EL10 EasyPack |
Pack of 1000 Each for $5.28
|
|
||
F1735001G
|
Gilson™
F1735001 |
200, 500, 1000μL | 10.22cm | 50 to 1250 μL | E1000 XL Tipack |
Pack of 384 Each for $43.08
|
|
||
F1736001G
|
Gilson™
F1736001 |
10, 50, 100μL | 5.857cm | 1 to 300 μL | E300 Tipack |
Pack of 768 Each for $73.84
|
|
||
F1736011G
|
Gilson™
F1736011 |
10, 50, 100μL | 5.857cm | 1 to 300 μL | E300ST Sterilized Tipack |
Pack of 768 Each for $83.06
|
|
||
F1739311G
|
Gilson™
F1739311 |
10, 50, 100μL | 5.144cm | 1 to 200 μL | E200 EasyPack |
Pack of 1000 Each for $33.84
|
|
||
Description
- Clear packaging allows easy identification of tips
- Color-coded tip packaging for volume color matching to pipettes
- Graduations tips enable quick-checks when aspirating samples
- Economical and easy-to-use
- Manufactured in sterile, clean-room environment
- Free of Human DNA, RNase, DNase, PCR Inhibitors and Pyrogens
Specifications
Standard Pipette Tips | |
Clear | |
EXPERT | |
Polypropylene |