missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ MEM Non-Essential Amino Acids Solution (100X) DFS Item

Used as a growth supplement for cell culture medium, to increase cell growth and viability

$31.38 - $544.00

Specifications

Product Type MEM Non-Essential Amino Acids Solution (100X)
View More Specs

Products 2 Description

Description

  • Contains the same non-essential amino acids found in the standard Minimum Essential Medium (MEM) at a strength of 100X
  • For in vitro diagnostic use
  • Manufactured at a cGMP compliant facility, located in Grand Island, New York, registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

MEM Non-Essential Amino Acids Solution (100X)

For Research Use Only. Not for use in diagnostic procedures.