missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ HEPES (1M)
A zwitterionic organic chemical buffering agent commonly used in cell culture media
Supplier: Gibco™ 15630080
Description
- HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media
- The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator
- A good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable
- It also has very low visible and UV light absorbance
- Store at 2°C to 8°C
Specifications
Zwitterionic Organic Chemical Buffer | |
Chromatin biology, mammalian cell culture | |
100 mL | |
N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid | |
Biochemical |
Clear | |
Liquid | |
Buffer | |
Sterile-filtered |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.