missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, Qualified, USDA-approved Regions DFS Item

Trusted quality for consistent performance

$820.00 - $1,584.00

Specifications

Format Bottle
Grade Qualified
Endotoxin Concentration ≤50 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤25mg/dL
View More Specs

Products 2 Description

Description

  • Sera Category: Standard
  • Sera for cell culture with robust cell lines
  • Good value for basic research
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25mg/dL
  • Origin: Countries meeting USDA importation requirements

Gibco Serum Delivers

  • ISO 13485 certified, processed in FDA registered facilities
  • Labeled for in vitro diagnostics (compliant with highest level of USP sterility testing)
  • Triple filtered at 0.1μm
  • Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Bottle
≤50 EU/mL
≤25mg/dL
Qualified
No