missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, Qualified, Heat Inactivated, USDA-approved Regions DFS Item

General stem cell culture media and reagents; sera for specialty research and specific assays

$229.00 - $1,640.00

Specifications

Format Bottle
Grade Qualified
Endotoxin Concentration ≤ 50 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤25 mg/dL
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
10438034 View Documents Gibco™
LS10438034
1000mL Each for $1,640.00
10438018 View Documents Gibco™
LS10438018
100mL Each for $229.00
10438026 View Documents Gibco™
LS10438026
500mL Each for $872.00
Description

Description

  • Sera Category: Standard
  • Origin: Countries meeting USDA importation requirements
  • Endotoxin level: ≤50EU/mL (levels routinely ≤ 10EU/mL)
  • Hemoglobin level: ≤25mg/dL

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Bottle
≤ 50 EU/mL
≤25 mg/dL
Qualified
Yes

For In Vitro Diagnostic Use.