missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Fetal Bovine Serum, certified, heat inactivated, United States
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
$157.04 - $1472.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Country of Origin | United States |
Serum Treatment | Heat-inactivated |
Endotoxin Concentration | ≤5 EU/mL |
Hemoglobin Concentration | ≤15 mg/dL |
Description
- Sera Category: Performance Plus
- Origin: United States
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤10mg/dL
Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Heat-inactivated | |
≤15 mg/dL | |
Frozen |
United States | |
≤5 EU/mL | |
Certified |
For In Vitro Diagnostic Use.