Learn More
Gibco™ DPBS, no calcium, no magnesium
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
$50.50 - $1152.00
Specifications
Classification | Animal Origin-Free |
---|---|
Product Type | DPBS |
Solution Type | Dulbecco's Phosphate Buffered Saline |
For Use With (Application) | Mammalian Cell Culture |
Osmolality | 270 - 300 mOsm/kg |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
14190144
|
Gibco™
14190144 |
500 mL |
Each for $50.50
|
|
|||||
14190250
|
Gibco™
14190250 |
10 x 500 mL |
Each for $430.00
|
|
|||||
14190136
|
Gibco™
14190136 |
1000 mL |
Each for $83.75
|
|
|||||
14190235
|
Gibco™
14190235 |
6 x 1000 mL |
Each for $455.00
|
|
|||||
14190342
|
Gibco™
14190342 |
5 L |
Each for $476.00
|
|
|||||
14190367
|
Gibco™
14190367 |
20 L |
Each for $1,152.00
|
|
|||||
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Order Info
Shipping Condition: Room Temperature
Specifications
Animal Origin-Free | |
Dulbecco's Phosphate Buffered Saline | |
270 - 300 mOsm/kg | |
No Phenol Red | |
Liquid |
DPBS | |
Mammalian Cell Culture | |
7.0 to 7.3 | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.