missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ CD Hybridoma Medium
Specifically developed for the growth of hybridomas and myelomas and monoclonal antibody production
Supplier: Gibco™ 11279023
Description
CD Hybridoma Medium is an animal origin-free, chemically defined, protein-free medium.
- Ability to support suspension and stationary hybridomas and myelomas
- Formulated without L-glutamine for stability
- Animal origin-free, protein-free, and chemically defined formulation
Ability to support suspension and stationary hybridomas and myelomas
- Suitable for the culture of recombinant myeloma lines as well as traditional hybridomas, Gibco CD Hybridoma Medium provides high cell densities and IgG production
- Cholesterol-dependent cultures such as NS0 require the addition of 250X Cholesterol Lipid Concentrate
- Formulated without L-glutamine for stability
- Gibco CD Hybridoma Medium requires the addition of 8 mM L-glutamine or GlutaMAX™ supplement before use
- GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format
- Animal origin-free, protein-free, and chemically defined formulation
- Gibco CD Hybridoma Medium is animal origin-free, protein-free, and chemically defined, allowing for easier purification of your protein of interest
- Gibco chemically defined media contain no proteins, hydrolysates, or components of unknown composition
Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture
Order Info
Shipping Condition: Room temperature
Specifications
Storage conditions: 2-8°C. Protect from light Shipping conditions: Ambient Shelf life: 18 months from date of manufacture |
|
Animal Origin-free, Chemically-defined, Protein-free | |
Hybridoma Medium | |
Sustainable packaging | |
Room Temperature |
Myeloma Cells, Hybridomas | |
Liquid | |
No Glutamine, No Phenol Red | |
Gibco™ |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.