missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ GIANT-DIGITS™ Radio Atomic Traceable Clock with Humidity

Fisherbrand GIANT-DIGITS™ Radio Atomic Traceable Clock with Humidity allows users to view time, temperature and humidity from anywhere in the lab

Manufacturer:  Fisherbrand™ 1088*

Catalog No. 15079680



Includes: Batteries, Wall mount, Bench stand, Traceable Certificate

Description

Description

  • Radio signal from NIST's Colorado atomic clock automatically sets correct date/time
  • calendar displays month/date, day, alarm, desk stand and wall mount supplied, plus temperature and humidity
  • Giant-Digits™ 4-1/2 inches viewable from 200 feet
  • Temperature range: 0 to 45°C (32 to 113°F); resolution: 0.1°; accuracy: ±1°C
  • Humidity range: 20 to 90% RH; resolution: 0.1% RH; accuracy: ±5% RH mid-range (±7% RH otherwise)

Certifications

ISO 17025, A2LA, NIST

Warranty

1 year from date of purchase

Specifications

Specifications

Battery power
Time of Day Alarm
ISO 17025, A2LA, NIST
28.7cm
43.2cm
3.3cm
LCD
±5% mid-range (±7% otherwise)
AA batteries (4)
17 x 1.3 x 11.3in.
Batteries, Wall mount, Bench stand, Traceable Certificate
0.45kg
Time: 1/10,000 of a second
Wall Clock
Documents