Genlantis™ Porcine Endothelial Cells
Economical alternative to human endothelial cells
$1,815.27 - $1,875.94
Specifications
Animal Porcine
For Use With (Application) To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells
Quantity >500000 Cells
Storage 4°C
View More Specs
Products 3
Catalog Number Mfr. No. Cell Type Price Quantity  
Catalog Number Mfr. No. Cell Type Price Quantity  
PP30005 View Documents Genlantis
PP30005
PCAEC Each for $1,815.27
PP30205 View Documents Genlantis
PP30205
PPAEC Each for $1,875.94
PP30405 View Documents Genlantis
PP30405
PAOEC Each for $1,875.94
Description
Description
  • Each lot functionally characterized
  • Same quality standard as the PrimaPure™ human cell products
  • Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
  • Supports 16 population doublings (12 for PCAEC) with Genlantis™ media and reagents
Specifications
Specifications
Porcine
To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells
>500000 Cells
4°C