Genlantis™ NeuroPure™ E18 Primary Rat Cortical Astrocytes
Save time with fresh, ready-to-use cells
Manufacturer:  Genlantis N900200
Catalog No. N900200
Includes: Day 18 Embryonic Sprague/Dawley Rat Cortical Astrocytes (1 vial x 2mL); NeuroPure™ Astrocyte Plating Medium (1 vial x 12mL); NeuroPrep™ Medium (1 vial x 2.5mL); NeuroPapain™ Enzyme (1 vial x 5mg)
Description
Description
NeuroPure™ E18 Primary Rat Cortical Astrocytes are live cells provided as micro-surgically dissected regions of day 18 embryonic Sprague/Dawley rat brain. These cells are prepared fresh each week and shipped in a nutrient rich medium that keeps them alive for up to 14 days under refrigeration. Following simple dissociation steps, the NeuroPure cells can be quickly plated on almost any poly-lysine coated substrate using the provided NeuroPure Plating Medium.
  • Freshly isolated healthy cells—never frozen
  • Get your primary culture up and running within one hour
  • Pure neuronal cells, greater than 95% astrocytes
Transfection, Pharmacology, Immunocytochemistry, Electrophysiology, and Neuronal Development Studies.
Specifications
Specifications
Transfection, pharmacology, electrophysiology, immunocytochemistry and neuronal development studies
4° to 8°C
Cortical Astrocytes
Day 18 embryonic sprague/dawley Rat cortical astrocytes (1 vial x 2mL), NeuroPure™ astrocyte plating medium (1 vial x 12mL), NeuroPrep™ medium (1 vial x 2.5mL), NeuroPapain™ Enzyme (1 vial x 5mg)
Rat
2 Million Cells
Documents