Genlantis™ Bovine Smooth Muscle Cell Kits
Economical alternative to primary human smooth muscle cells
$2,418.18 - $2,432.53
Specifications
Animal Bovine
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
Includes Cryopreserved cells, growth medium, subculture reagent kit
Quantity >500000 Cells
Storage 4°C
View More Specs
Products 3
Catalog Number Mfr. No. Cell Type Price Quantity  
Catalog Number Mfr. No. Cell Type Price Quantity  
PB35005K View Documents Genlantis
PB35005K
BCASMC Each for $2,418.18
PB35205K View Documents Genlantis
PB35205K
BPASMC Each for $2,418.18
PB35405K View Documents Genlantis
PB35405K
BAOSMC Each for $2,432.53
Specifications
Specifications
Bovine
Cryopreserved cells, growth medium, subculture reagent kit
4°C
Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched bovine arterial endothelial cells
>500000 Cells