Genlantis™ Bovine Endothelial Cell Complete Kits
Economical alternative to human endothelial cells
Supplier: Genlantis™ PB30005
Description
- Each lot functionally characterized
- Same quality standard as the PrimaPure human cell products
- Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
- 16 population doublingsguaranteed with Genlantis media and reagents
- Economical alternative suitable for the studies of endothelial function and endothelial metabolism, especially in co-culture of species-matched bovine arterial smooth muscle cells
Specifications
4°C | |
Bovine | |
— |
Coronary Artery (BCAEC) | |
To study endothelial function and endothelial metabolism, especially in co-culture of species matched bovine arterial smooth muscle cells | |
>500000 Cells |