Genlantis™ Bovine Endothelial Cell Complete Kits
Economical alternative to human endothelial cells
$1,735.53 - $2,242.76
Specifications
For Use With (Application) To study endothelial function and endothelial metabolism, especially in co-culture of species matched bovine arterial smooth muscle cells
Storage 4°C
View More Specs
Products 6
Catalog Number Mfr. No. Animal Cell Type Includes Quantity Price Quantity  
Catalog Number Mfr. No. Animal Cell Type Includes Quantity Price Quantity  
PB30005 View Documents Genlantis
PB30005
Bovine Coronary Artery (BCAEC) >500000 Cells Each for $1,744.02
PB30005K View Documents Genlantis
PB30005K
Bovine Coronary Artery (BCAEC), Complete Kit Cryopreserved cells, growth medium, subculture reagent kit >500000 Cells Each for $2,240.57
PB30205 View Documents Genlantis
PB30205
Bovine Pulmonary Artery (BPAEC) >500000 Cells Each for $1,740.63
PB30205K View Documents Genlantis
PB30205K
Bovine Pulmonary Artery (BPAEC),Complete Kit Cryopreserved cells, growth medium, subculture reagent kit >500000 Cells Each for $2,238.39
PB30405K View Documents Genlantis
PB30405K
Bovine Aortic Cella (BAOEC), Complete Kit Cryopreserved cells, growth medium, subculture reagent kit >500000 Cells Each for $2,242.76
PB30405 View Documents Genlantis
PB30405
Bovine Aortic Cella (BAOEC) >500000 Cells Each for $1,735.53
Description
Description
  • Each lot functionally characterized
  • Same quality standard as the PrimaPure human cell products
  • Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
  • 16 population doublingsguaranteed with Genlantis media and reagents
  • Economical alternative suitable for the studies of endothelial function and endothelial metabolism, especially in co-culture of species-matched bovine arterial smooth muscle cells
Specifications
Specifications
To study endothelial function and endothelial metabolism, especially in co-culture of species matched bovine arterial smooth muscle cells
4°C