missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ BioCooler™ Cold Box
Convenient benchtop cooler for optimal enzyme stability
Supplier: Genlantis™ E100100
Description
- Constant temperature of -15°C or below for up to 4 hours on the benchtop
- Capacity for 48 vials (1.5 to 2.0mL each)
- Textured outer surfaces and carrying handle for ease of transport
- Hinged and locked lid for maximum security during handling
- Erasable interior note panel for easy sample identification
- Black exterior, white tube rack
Order Info
L x W x H: 8.5 x 3.5 x 8 in. (21.5 x 9.5 x 20 cm); Weight: 2.5kg
Specifications
20 cm | |
21.5 cm | |
8.9 cm | |
8.5 x 3.5 x 8 in. (21.5 x 8.9 x 20 cm) | |
5.51 lb. | |
BioCooler Cold Box | |
48 (1.5 to 2mL vials at -15°C or below for up to 4 hours) |
8 in. | |
8.5 in. | |
3.5 in. | |
Hinged Locked Lid | |
2.4 kg | |
Black, White | |
-15°C |