missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Dextrin Sepharose™ High Performance Media
Provide convenient affinity purification of recombinant proteins with maltose binding protein (MBP) tags
$1666.86 - $5316.72
Specifications
Material | Dextrin Sepharose |
---|---|
Particle Size | 34 μm |
dimMatrix | Affinity Resin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
45-001-583
|
Cytiva
28935597 |
25 mL |
Each for $1,666.86
|
|
|||||
45-001-584
|
Cytiva
28935598 |
100 mL |
Each for $5,316.72
|
|
|||||
Description
- Physiological conditions and mild elution preserves activity of target protein
- Robust, stable medium allows narrow peaks in elution of target protein
- Compatible with commonly used aqueous buffers and easily regenerated with 0.5M NaOH
- Easy scale-up
Specifications
Dextrin Sepharose | |
34 μm | |
Affinity Resin |