missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Formalin, Buffered, 10% (Phosphate Buffer/Certified), Fisher Chemical™
$292.92 - $15386.17
Specifications
Chemical Name or Material | Formalin, Buffered, 10% |
---|---|
Physical Form | Liquid |
Melting Point | 0°C |
Name Note | Phosphate Buffer |
Boiling Point | 93.9°C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
SF100-4
|
Fisher Chemical
SF1004 |
4 L | Poly Bottle |
Each for $292.92
Case of 4 Each for $1,171.68
|
|
||||
SF100-20
|
Fisher Chemical
SF10020 |
20 L | PolyPac™ |
Each for $807.50
|
|
||||
SF100-200
|
Fisher Chemical
SF100200 |
200 L | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
Formalin, Buffered, 10% | |
0°C | |
93.9°C | |
1.10g/cm³ | |
Specially filtered for use in tissue processors | |
MFCD00003274 | |
Certified |
Liquid | |
Phosphate Buffer | |
50-00-0,67-56-1,7558-79-4,10049-21-5,7732-18-5 | |
6.9 to 7.1 | |
Assay: 4 to 5% w/w | |
4 to 5% | |
10% w/v (Phosphate Buffer) |