missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Forma™ Flammable Storage Freezer, -20°C; 23 cu. ft., 115V

Protect flammable materials, solvents and intermediates using Thermo Scientific™ Forma™ Flammable Materials Freezers.

Manufacturer:  Thermo Scientific FFMS2320A

Catalog No. FFMS2320A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Protect flammable materials, solvents and intermediates using high-performance Thermo Scientific™ Forma™ Flammable Materials Freezers. Designed to meet stringent U.S. Pharmacopeia standards for drug storage and  NFPA safety provisions for storing flammable liquids.† These freezers are designed to maintain a safe environment while keeping valuable samples within a tight temperature range. 


Each is listed by UL as a special purpose freezer intended for flammable material storage in accordance with NFPA 45, 70 and 99.† 45, 70 and 99.†

Key Features:
  • Easy to use intuitive digital display featuring microprocessor control
  • Lockable doors, audio and visual alarms including door ajar alarms for security and peace of mind
  • Convenient access port and casters
  • Outstanding construction featuring painted, high-quality steel exteriors with durable finish
  • Ignition-free interior cabinet design for the safe storage of flammable materials
  • Meets safety standards in accordance with UL, NFPA† and OSHA† for regulatory compliance
  • Optional, built-in, inkless, seven-day graphic chart recorder
  • Optional, barrier-protected 4-20 milliamp output for external monitoring
  • Manual defrost

Ordering Information:

Temperature chart recorders, and 4-20 milliamp are available separately.

Note:

† Products complying with the applicable requirements of NFPA 70, “National Electric Code”, NPFA 45, “Fire Protection for Laboratories Using Chemicals,” and NFPA 99 “Health Care Facilities” are considered to meet the OSHA guidelines for these types of facilities. These units have no internal electrical components that could trigger an explosion or fire of flammable materials inside the unit. Flammable material storage units are not for use in classified hazardous locations which require the use of explosion-proof units.

Specifications

Specifications

13.5A
Scratch-resistant painted steel
73.7cm
58 in.
147.3cm
CFC-free
28 in.
72cm
Manual
23.3 cu. ft.
37 in.
29 x 24 x 58 in. (73.7 x 61 x 147.3cm)
115V
20A
29 in.
78.6 in.
200cm
60Hz
4
24 in.
61cm
UL and cUL listed, CE marked
659L
94cm
−20°C
115V 60Hz
Documents