Thermo Scientific™ Forma™ Environmental Chambers
Perform a variety of applications with these environmental chambers, which can be configured for a variety of purposes and are available in multiple capacities.
Specifications
Certifications/Compliance ICH, FDA, TAPPI, ASTM™
Compressor 1/4hp, air-cooled
Defrost Adjustable defrost timer; factory set for two 15-min. defrost cycles/24 hr.
Cabinet Material Cold-rolled steel, powder-coated cabinet
Display Digital dual LED
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (English) Voltage Price Quantity  
Catalog Number Mfr. No. Capacity (English) Voltage Price Quantity  
13-987-065 View Documents Thermo Scientific™
FOR3911*
11 cu. ft. 208/220V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-262-340 View Documents Thermo Scientific™
FOR3920
29 cu. ft. 120V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13-067-066 View Documents Thermo Scientific™
FOR3940
29 cu. ft. 208/220V N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Perform a variety of applications withThermo Scientific™ Forma™ environmental chambers, which can be configured for a variety of purposes and are available in multiple capacities. With a wide range of accessories and years of experience in constructing cabinets, we can construct units to match your most stringent applications.
Broad Range of Features and Controls:
  • Wide temperature range of 0° to 60°C that enhances the flexibility of the unit
  • Relative humidity control via steam generator from above ambient to 95% at 37°C (not available w/Cat. No. 3920)
  • Sophisticated digital controls
  • Programmable ramp and soak cycles for temperature and RH†
  • Durable cabinets with true laminar airflow through side plenums for extremely tight uniformity
  • Phenolic-coated evaporator for use in applications that create corrosive byproducts
  • Manual control of defrost and refrigeration cycles
  • Visible/audible safety alarms alert to over- and undertemperature conditions, deviation of humidity from set parameters (except Model 3920)
Durable Construction:
  • Cold-rolled steel, powder-coated cabinet
  • 2″ (5.1cm) thick, CFC-free, foamed-in-place polyurethane insulation
  • Stainless-steel interior with solid stainless-steel adjustable shelves
  • Directed horizontal airflow system for excellent uniformity
  • Locking swivel casters for easy mobility during installation and cleaning
  • Leveling feet for stability and added safety in the lab
Broad Range of Options:
  • Interior lighting for Models 3920, 3940 and 3949 (Optional)
  • Reinforced shelving and flooring for heavy loads
  • Heatless dryer for lower humidity levels
  • Access ports for probes, sensors
  • Interior convenience receptacles
  • CO2 (IR sensor)
  • 6″ and 12″ temperature chart recorder
  • Other custom options available
Compatible with:
  • Recorders (197075, 198036, 198039, 198042, 201144, 201145, 201146, 201147)
  • Reach-In/Environmental Chamber Solid Stainless-steel Shelf (224139)
  • Reach-In/Environmental Chamber Perforated Stainless-steel Shelf (224155)
  • Reinforced Stainless-steel Shelf (224161)
  • Fixed Reinforced Stainless-steel Shelf for two shakers (1900005)
  • Stainless-steel Door Glass Cover, factory installed (190591)
Recommended for:
Ideal for growth, stability and shelf-life testing applications
Ordering Information
† Humidity controller available on 3940/3949, 3911/3913 only
Specifications
Specifications
ICH, FDA, TAPPI, ASTM™
Adjustable defrost timer; factory set for two 15-min. defrost cycles/24 hr.
Digital dual LED
Temperature ramp and soak, up to 24 steps
±0.1 at 25° to 37°C
0.1°C
±0.3 at 25° to 37°C
1/4hp, air-cooled
Cold-rolled steel, powder-coated cabinet
1750W (6000 BTU/hr.) at 220V, 1600W (5500BTUH) at
Relative humidity control via steam generator from above ambient to 95% at 37°C
0° to 60°C
RTD
CFC-free, R134A