missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ FLoid™ Cell Imaging Station DFS Item

Affordable, user-friendly imaging solution for quick detection and verification of fluorescently-labeled samples

Manufacturer:  Invitrogen™ LS4471136

Catalog No. 4471136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Uses an open-stage design providing maximum vessel flexibility for slides, coverslips, multi-well culture dishes, or even T75 flasks. Captures transmitted light and three-color fluorescent images of cells and samples at benchtop without special training or dedicated staff. Simple user interface, streamlined image acquisition process, and real-time, multicolor display allow users to produce high-quality images with a few mouse clicks.

  • Simplicity: Fully integrated system with intuitive, multilingual user interface
  • Speed: Get results in a snap, without warm-up, cool-down, or filter changes
  • Convenience: Capture and print images on your bench, rather than in the darkroom
  • Information: Provides information on Molecular Probes™ reagents and protocols via user interface
  • Robustness: No moving parts and long life LEDs for reliable day to day use
  • High Quality Imaging Anywhere in Lab
  • Versatile and easy to use
  • Fast and reliable
  • An everyday use imager for every user

System specifications

  • Objective: Fixed 20X plan fluorite; numerical aperture (NA)= 0.45; working distance (WD)=5.9 mm
  • Color Channels: 4 channels (relief phase, blue, green, and red fluorescence)
  • Illumination: LED (50,000 hour life), adjustable intensity
  • Excitation: Blue channel: 390/40nm; Green channel: 482/18nm; Red channel: 586/15nm
  • Emission: Blue channel: 446/33nm; Green channel: 532/59nm; Red channel: 646/68nm
  • Captured Images: 16-bit monochrome, TIFF, PNG, JPG, or BMP (129X 964 pixels)
  • LCD display: 15 in. color; display resolution 1366 x 768 pixels; image resolution 1296 x 964 pixels; adjustable tilt

Cell Analysis, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

W x H x D: (40 x 54 x 35cm)16 x 21 x 14 in. Weight: 12.7kg (28 lb.). Shipping Condition: Room Temperature.

Specifications

Specifications

54cm
Cell Imaging Station
35cm
40cm
21 in.
14 in.
16 in.
Documents