missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BUCHI FlashPure Select Flash Chromatography Cartridges: C18
FlashPure Select cartridges are designed for ultimate and unmatched performance.
$484.30 - $1858.57
Specifications
Material | Polypropylene |
---|---|
Type | C18 |
Resolution | 20 to 35 μm |
Column Type | C18 |
For Use With (Application) | Non-Polar Compounds, Ideal for Peptides |
Catalog Number | Mfr. No. | Size (Metric) Tube Rack | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (Metric) Tube Rack | Includes | Price | Quantity | ||||
05410092
|
BUCHI
140000126 |
120 g | 2 Pc. |
Pack of 2 Each for $1,476.48
|
|
||||
05410088
|
BUCHI
140000122 |
12 g | 5 Pc. |
Pack of 5 Each for $1,128.98
|
|
||||
05410089
|
BUCHI
140000123 |
20 g | 2 Pc. |
Pack of 2 Each for $484.30
|
|
||||
05410093
|
BUCHI
140000127 |
220 g | 1 Pc. |
Pack for $1,520.50
|
|
||||
05410094
|
BUCHI
140000128 |
330 g | 1 Pc. |
Pack for $1,858.57
|
|
||||
05410090
|
BUCHI
140000124 |
40 g | 2 Pc. |
Pack of 2 Each for $580.21
|
|
||||
05410087
|
BUCHI
140000121 |
4 g | 5 Pc. |
Pack of 5 Each for $610.09
|
|
||||
05410091
|
BUCHI
140000125 |
80 g | 2 Pc. |
Pack of 2 Each for $1,069.23
|
|
||||
Description
- Spherical particles
- 13-17 μm silica, 20-35 μm C18
- Functionalized silica cartridges can be reused multiple times
- Large selection of cartridge sizes from 4 g - 3,000 g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen.
Specifications
Polypropylene | |
20 to 35 μm | |
Non-Polar Compounds, Ideal for Peptides |
C18 | |
C18 | |
Any Flash System |