Fisherbrand™ Universal Mini Economy Spill Kit
Ideal for quick cleanup of small drips and spills.
$8.92 - $63.18
Specifications
Certifications/Compliance | NSF, TCLP |
---|---|
For Use With (Application) | Quick cleanup of small drips and spills |
Alert:
Not suitable for use with hydrofluoric acid.
Includes: One carton of super sorbent; one pair each of goggles and nitrile gloves; one dustpan/broom; one disposal bag; universal instruction sheet.
Description
- Six individually packaged kits per box
- Each kit absorbs 0.6 gal. (2.3L)
- Lightweight, high-capacity particulate sorbent (also available separately) is nonbiodegradable, inert, noncorrosive, nontoxic and nonflammable
- Instantly absorbs oils, acids (except hydrofluoric acid), bases, chemicals and toxic liquids
Certifications
Sorbent is National Sanitation Foundation (NSF) certified
Compliance
Sorbent exceeds landfill rules and passes Paint Filter and TCLP tests
Specifications
NSF, TCLP | |
Quick cleanup of small drips and spills |