missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Superfrost™ Disposable Microscope Slides
Superfrost coating impervious to most common laboratory chemicals and reagents
$83.09 - $121.33
Specifications
Frosted | SuperFrost |
---|---|
Type | Disposable Microscope Slide |
Width (Metric) | 25 mm |
Color | White |
Coating | Opaque |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
12-550-123
|
Fisherbrand™
4951F602811T |
Superfrost White Tab Slide, Full Size |
N/A
|
|
|||||
12-550-133
|
Fisherbrand™
4955F600621T |
Superfrost Slide, 1.2mm thick |
N/A
|
|
|||||
12-550-143
|
Fisherbrand™
4951602811T |
Superfrost White Tab Slide, Standard Metric Size Slide |
N/A
|
|
|||||
Description
Superfrost coating impervious to most common laboratory chemicals and reagents
- Brilliant white, opaque coating takes and retains most laboratory marking methods
- Each side has a written key for easy orientation; the word “SPECIMEN” instantly tells you which side is up
- Slides can be imprinted with your laboratory or hospital's name (call your Fisher HealthCare™ Sales Representative for details)
Order Info
Come in handy one-at-a-time dispenser boxes of 72 slides.
Specifications
SuperFrost | |
25 mm | |
Opaque | |
1 in. |
Disposable Microscope Slide | |
White | |
3 in. | |
3 x 1 in. (75 x 25 mm) |