Fisherbrand™ Serialized Class A Cylinders with Certificate of Traceability
Calibrated “To Deliver”
$131.00 - $589.07
Specifications
Base Type | Hexagonal |
---|---|
Calibration | To Deliver |
Class | A |
Type | Graduated Cylinder |
Scale Color | White |
Includes: Certificate that indicates cylinder is individually calibrated and certified against equipment whose calibration is traceable to the National Institute of Standards and Technology (NIST™); lists the weight set used, the test number, and the date of testing
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Tolerance Metric | Subdivision | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Graduations | Tolerance Metric | Subdivision | Price | Quantity | ||
08-557C
|
Fisherbrand™
08557C |
50 mL | 1 mL | ±0.25 mL | 1mL |
Each for $131.00
Case of 6 Each for $786.00
|
|
||
08-557B
|
Fisherbrand™
08557B |
25 mL | 0.2 mL | ±0.17 mL | 0.2mL |
Each for $154.47
Case of 6 Each for $926.82
|
|
||
08-557A
|
Fisherbrand™
08557A |
10 mL | 0.1 mL | ±0.1 mL | 0.1mL |
Each for $132.37
Case of 6 Each for $794.22
|
|
||
08-557D
|
Fisherbrand™
08557D |
100 mL | 1 mL | ±0.5 mL | 1mL |
Each for $137.97
Case of 6 Each for $827.82
|
|
||
08-557E
|
Fisherbrand™
08557E |
250 mL | 2 mL | ±1 mL | 2mL |
Each for $187.33
Case of 4 Each for $749.32
|
|
||
08-557H
|
Fisherbrand™
08557H |
2000 mL | 20 mL | ±6 mL | 20mL |
Each for $589.07
|
|
||
08-557F
|
Fisherbrand™
08557F |
500 mL | 5 mL | ±2 mL | 5mL |
Each for $237.83
Case of 4 Each for $951.32
|
|
||
09-730-30
|
Fisherbrand™
08557G |
1000 mL | 10 mL | ±3 mL | 10mL |
Each for $300.70
Case of 2 Each for $601.40
|
|
||
Description
Calibrated “To Deliver”
- Serialized
- White scale
- Molded hexagonal base
Specifications
Hexagonal | |
A | |
White | |
Borosilicate Glass | |
White scale graduations |
To Deliver | |
Graduated Cylinder | |
Round | |
Traceable NIST Certificate |