missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Robotic Screw Top Autosampler Vial Closures, Closure with PTFE/Silicone Septa

For use with Fisherbrand™ Robotic Screw Top Autosampler Vials (cat. no. 033918).

$32.80 - $51.07

Specifications

Closure Type Screw top
Color Blue
Vial Closure Size 9mm
View More Specs

Products 3 Description

Description

For use with Fisherbrand™ Robotic Screw Top Autosampler Vials (cat. no. 033918).

  • Blue closures are preassembled with a 9mm thread and choice of PTFE/Silicone, PTFE/Silicone/PTFE, or PTFE/Red Rubber septa

Specifications

Specifications

Screw top
Blue
9mm